Anti-MTOR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MTOR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-607
- 77855-605
- Antigen Symbol:
- MTOR
- Antigen Synonyms:
- dJ576K7.1 (FK506 binding protein 12 rapamycin associated protein 1)|FK506 binding protein 12 rapamycin associated protein 1|FK506 binding protein 12 rapamycin associated protein 2|FK506 binding protein 12 rapamycin complex associated protein 1|FK506-binding protein 12-rapamycin complex-associated protein 1|FKBP rapamycin associated protein|FKBP12 rapamycin complex associated protein|FKBP12-rapamycin complex-associated protein 1|FKBP12-rapamycin complex-associated protein|FLJ44809|FRAP|FRAP1|FRAP2|Mammalian target of rapamycin|Mechanistic target of rapamycin|mTOR|MTOR_HUMAN|OTTHUMP00000001983|RAFT1|Rapamycin and FKBP12 target 1|Rapamycin associated protein FRAP2|Rapamycin target protein 1|Rapamycin target protein|RAPT1|Serine/threonine-protein kinase mTOR
- dJ576K7.1 (FK506 binding protein 12 rapamycin associated protein 1)|FK506 binding protein 12 rapamycin associated protein 1|FK506 binding protein 12 rapamycin associated protein 2|FK506 binding protein 12 rapamycin complex associated protein 1|FK506-binding protein 12-rapamycin complex-associated protein 1|FKBP rapamycin associated protein|FKBP12 rapamycin complex associated protein|FKBP12-rapamycin complex-associated protein 1|FKBP12-rapamycin complex-associated protein|FLJ44809|FRAP|FRAP1|FRAP2|Mammalian target of rapamycin|Mechanistic target of rapamycin|mTOR|MTOR_HUMAN|OTTHUMP00000001983|RAFT1|Rapamycin and FKBP12 target 1|Rapamycin associated protein FRAP2|Rapamycin target protein 1|Rapamycin target protein|RAPT1|Serine/threonine-protein kinase mTOR
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P42345
- P42345
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase mTOR protein (1223-1290AA)
- Recombinant Human Serine/threonine-protein kinase mTOR protein (1223-1290AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together