Anti-HCFC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HCFC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-779
- 77739-491
- Antigen Symbol:
- HCFC1
- HCFC1
- Antigen Synonyms:
- C1 factor|CFF|HCF 1|HCF|HCF C-terminal chain 6|HCF-1|HCF1|HCFC1_HUMAN|HFC1|Host cell factor 1|Host cell factor|Host cell factor C1 (VP16 accessory protein)|Host cell factor C1|MGC70925|MRX3|VCAF|VP16 accessory protein
- C1 factor|CFF|HCF 1|HCF|HCF C-terminal chain 6|HCF-1|HCF1|HCFC1_HUMAN|HFC1|Host cell factor 1|Host cell factor|Host cell factor C1 (VP16 accessory protein)|Host cell factor C1|MGC70925|MRX3|VCAF|VP16 accessory protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51610
- P51610
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Host cell factor 1 protein (201-428AA)
- Recombinant Human Host cell factor 1 protein (201-428AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together