Anti-LAIR1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LAIR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-949
- 77858-808
- Antigen Symbol:
- LAIR1
- LAIR1
- Antigen Synonyms:
- 5133400O11Rik|CD305|D7Bwg0421e|hLAIR 1|LAIR-1|LAIR1 protein|LAIR1_HUMAN|Leukocyte associated Ig like receptor 1|Leukocyte associated immunoglobulin like receptor 1|Leukocyte-associated immunoglobulin-like receptor 1
- 5133400O11Rik|CD305|D7Bwg0421e|hLAIR 1|LAIR-1|LAIR1 protein|LAIR1_HUMAN|Leukocyte associated Ig like receptor 1|Leukocyte associated immunoglobulin like receptor 1|Leukocyte-associated immunoglobulin-like receptor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6GTX8
- Q6GTX8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leukocyte-associated immunoglobulin-like receptor 1 protein (22-165AA)
- Recombinant Human Leukocyte-associated immunoglobulin-like receptor 1 protein (22-165AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together