Anti-HSPB11 Rabbit Polyclonal Antibody
Catalog # 77740-487
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HSPB11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9Y547
- Antigen Synonyms:Chromosome 1 open reading frame 41|Heat shock protein beta-11|HSB11_HUMAN|Hspb11|HSPCO34|IFT25|Placental protein 25|PP25
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:PP25
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HSPB11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat