Anti-MASP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MASP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-803
- 77859-502
- Antigen Symbol:
- MASP1
- MASP1
- Antigen Synonyms:
- CRARF|CRARF1|PRSS5Mannan-binding lectin serine protease 1|EC 3.4.21.-|Complement factor MASP-3|Complement-activating component of Ra-reactive factor|Mannose-binding lectin-associated serine protease 1|MASP-1|Mannose-binding protein-associated serine protease|Ra-reactive factor serine protease p100|RaRF|Serine protease 5) [Cleaved into: Mannan-binding lectin serine protease 1 heavy chain|Mannan-binding lectin serine protease 1 light chain]
- CRARF|CRARF1|PRSS5Mannan-binding lectin serine protease 1|EC 3.4.21.-|Complement factor MASP-3|Complement-activating component of Ra-reactive factor|Mannose-binding lectin-associated serine protease 1|MASP-1|Mannose-binding protein-associated serine protease|Ra-reactive factor serine protease p100|RaRF|Serine protease 5) [Cleaved into: Mannan-binding lectin serine protease 1 heavy chain|Mannan-binding lectin serine protease 1 light chain]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48740
- P48740
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mannan-binding lectin serine protease 1 protein (20-699AA)
- Recombinant Human Mannan-binding lectin serine protease 1 protein (20-699AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together