Anti-SPINK6 Rabbit Polyclonal Antibody
77738-966
New Product
- Antigen Symbol:SPINK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q6UWN8
- Antigen Synonyms:Acrosin inhibitor|BUSI2|EG433180|ISK6_HUMAN|MGC21394|Protease inhibitor H|Serine protease inhibitor Kazal-type 6|Spink5l1|SPINK6|UNQ844|UNQ844/PRO1782
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Acrosin inhibitor
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: SPINK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human