Anti-NQO2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NQO2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-890
- 77744-891
- Antigen Symbol:
- NQO2
- NQO2
- Antigen Synonyms:
- DHQV|DIA6|EC 1.10.99.2|MGC94180|NAD(P)H dehydrogenase quinone 2|NAD(P)H menadione oxidoreductase 1 dioxin inducible 2|NAD(P)H menadione oxidoreductase 2 dioxin inducible|NMOR2|NQO 2|NQO2_HUMAN|NRH dehydrogenase [quinone] 2|NRH dehydrogenase|NRH:quinone oxidoreductase 2|OTTHUMP00000015948|OTTHUMP00000015949|OTTHUMP00000015953|Ox 2|Ox2|QR2|Quinone|Quinone reductase 2|Ribosyldihydronicotinamide dehydrogenase [quinone]|Ribosyldihydronicotinamide dehydrogenase
- DHQV|DIA6|EC 1.10.99.2|MGC94180|NAD(P)H dehydrogenase quinone 2|NAD(P)H menadione oxidoreductase 1 dioxin inducible 2|NAD(P)H menadione oxidoreductase 2 dioxin inducible|NMOR2|NQO 2|NQO2_HUMAN|NRH dehydrogenase [quinone] 2|NRH dehydrogenase|NRH:quinone oxidoreductase 2|OTTHUMP00000015948|OTTHUMP00000015949|OTTHUMP00000015953|Ox 2|Ox2|QR2|Quinone|Quinone reductase 2|Ribosyldihydronicotinamide dehydrogenase [quinone]|Ribosyldihydronicotinamide dehydrogenase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16083
- P16083
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Ribosyldihydronicotinamide dehydrogenase [quinone] protein (1-231AA)
- Ribosyldihydronicotinamide dehydrogenase [quinone] protein (1-231AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together