Anti-MAP2K6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: MAP2K6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-650
- 77859-378
- Antigen Symbol:
- MAP2K6
- MAP2K6
- Antigen Synonyms:
- Dual specificity mitogen activated protein kinase kinase 6|Dual specificity mitogen-activated protein kinase kinase 6|MAP kinase kinase 6|MAPK/ERK kinase 6|MAPKK 6|MAPKK6|MEK 6|MEK6|Mitogen Activated Protein Kinase Kinase 6|MKK 6|MKK6|MP2K6_HUMAN|PRKMK6|protein kinase|Protein kinase mitogen activated kinase 6|protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6)|SAPK kinase 3|SAPKK-3|SAPKK3|SKK3|Stress-activated protein kinase kinase 3
- Dual specificity mitogen activated protein kinase kinase 6|Dual specificity mitogen-activated protein kinase kinase 6|MAP kinase kinase 6|MAPK/ERK kinase 6|MAPKK 6|MAPKK6|MEK 6|MEK6|Mitogen Activated Protein Kinase Kinase 6|MKK 6|MKK6|MP2K6_HUMAN|PRKMK6|protein kinase|Protein kinase mitogen activated kinase 6|protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6)|SAPK kinase 3|SAPKK-3|SAPKK3|SKK3|Stress-activated protein kinase kinase 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52564
- P52564
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 6 protein (1-334AA)
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 6 protein (1-334AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together