Anti-HRH1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HRH1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-382
- 77857-650
- Antigen Symbol:
- HRH1
- HRH1
- Antigen Synonyms:
- BPHS, mouse, homolog of|H1 R|H1R|HH1R|HisH1|Histamine H(1) receptor|Histamine H1 receptor|Histamine receptor H1|Histamine receptor subclass H1|Hrh1|HRH1_HUMAN
- BPHS, mouse, homolog of| H1 R| H1R| HH1R| HisH1| Histamine H(1) receptor| Histamine H1 receptor| Histamine receptor H1| Histamine receptor subclass H1| Hrh1| HRH1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35367
- P35367
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histamine H1 receptor protein (211-416AA)
- Recombinant Human Histamine H1 receptor protein (211-416AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together