Anti-OR10G9 Rabbit Polyclonal Antibody
77736-642
New Product
- Antigen Symbol:OR10G9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q8NGN4
- Antigen Synonyms:O10G9_HUMAN|Olfactory receptor 10G10|Olfactory receptor 10G9|olfactory receptor OR11-272|olfactory receptor|family 10|subfamily G|member 9|OR10G10P|OR10G9
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:O10G9_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IF:1:200 to 1:1000, ELISA:1:40000
Type:
Antigen: OR10G9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human