Anti-HUS1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: HUS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77857-773
- 77740-541
- Antigen Symbol:
- HUS1
- HUS1
- Antigen Synonyms:
- Checkpoint protein hHUS1 (S. pombe) checkpoint homolog|HUS1 checkpoint homolog (S. pombe)|HUS1 Checkpoint Protein|HUS1 protein|HUS1+ - like protein|HUS1_HUMAN|Hydroxyurea-sensitive 1, S. pombe, homolog of
- Checkpoint protein hHUS1 (S. pombe) checkpoint homolog|HUS1 checkpoint homolog (S. pombe)|HUS1 Checkpoint Protein|HUS1 protein|HUS1+ - like protein|HUS1_HUMAN|Hydroxyurea-sensitive 1, S. pombe, homolog of
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60921
- O60921
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Checkpoint protein HUS1 protein (2-280AA)
- Recombinant Human Checkpoint protein HUS1 protein (2-280AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together