Anti-NUP160 Rabbit Polyclonal Antibody
Catalog # 77745-213
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NUP160
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q12769
- Antigen Synonyms:160 kDa nucleoporin|DKFZp686M14102|FLJ22583|KIAA0197|MGC150678|MGC150679|NU160_HUMAN|Nuclear pore complex protein Nucleoporin 160kD|Nucleoporin 160kDa|Nucleoporin NUP 160|NUP120|Nup160
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:160 kDa nucleoporin
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NUP160
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat