Anti-EXOC7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EXOC7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-000
- 77736-286
- Antigen Symbol:
- EXOC7
- EXOC7
- Antigen Synonyms:
- 2 5 3p|DKFZp686J04253|EX070|EXO70|Exo70p|EXOC 1|EXOC 7|EXOC1|EXOC7_HUMAN|Exocyst complex component 7|Exocyst complex component Exo70|FLJ40965|FLJ46415|KIAA1067|Rexo70|YJL085W
- EXO70|EX070|EXOC 1|EXOC 7|KIAA1067|DKFZp686J04253|Exocyst complex component Exo70|YJL085W|Exo70p|Rexo70|FLJ46415|2 5 3p|FLJ40965|EXOC1|EXOC7_HUMAN|Exocyst complex component 7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPT5
- Q9UPT5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Exocyst complex component 7 protein (521-735AA)
- Recombinant Human Exocyst complex component 7 protein (521-735AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together