Anti-ACVRL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACVRL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-825
- 77855-189
- Antigen Symbol:
- ACVRL1
- ACVRL1
- Antigen Synonyms:
- Activin A receptor|Activin A receptor type II like 1|type II like kinase 1|Activin receptor like kinase 1|Activin receptor-like kinase 1|ACVL1_HUMAN|ACVRLK1|ALK-1|ALK1|HHT|HHT2|ORW2|Osler Rendu Weber syndrome 2|Serine/threonine protein kinase receptor R3|Serine/threonine-protein kinase receptor R3|SKR3|TGF B superfamily receptor type I|TGF-B superfamily receptor type I|TSR-I|TSR1
- Activin A receptor|Activin A receptor type II like 1|type II like kinase 1|Activin receptor like kinase 1|Activin receptor-like kinase 1|ACVL1_HUMAN|ACVRLK1|ALK-1|ALK1|HHT|HHT2|ORW2|Osler Rendu Weber syndrome 2|Serine/threonine protein kinase receptor R3|Serine/threonine-protein kinase receptor R3|SKR3|TGF B superfamily receptor type I|TGF-B superfamily receptor type I|TSR-I|TSR1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P37023
- P37023
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase receptor R3 protein (22-118AA)
- Recombinant Human Serine/threonine-protein kinase receptor R3 protein (22-118AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together