Anti-IRS1 Rabbit Polyclonal Antibody
77741-219
New Product
- Antigen Symbol:IRS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P35568
- Antigen Synonyms:HIRS 1|HInsulin receptor substrate 1|IRS 1|IRS-1|IRS1_HUMAN|OTTHUMP00000164234
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:OTTHUMP00000164234
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat