Anti-NDUFB6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFB6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-195
- 77744-196
- Antigen Symbol:
- NDUFB6
- NDUFB6
- Antigen Synonyms:
- B17|CI|CI B17|CI-B17|Complex I mitochondrial respiratory chain B17 subunit|Complex I-B17|Complex IB17|MGC13675|NADH dehydrogenase (ubiquinone) 1 beta subcomplex subunit 6|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6|NADH ubiquinone oxidoreductase B17 subunit|NADH ubiquinone oxidoreductase beta subunit 6|NADH-ubiquinone oxidoreductase B17 subunit|NDUB6_HUMAN|NDUFB6
- B17|CI|CI B17|CI-B17|Complex I mitochondrial respiratory chain B17 subunit|Complex I-B17|Complex IB17|MGC13675|NADH dehydrogenase (ubiquinone) 1 beta subcomplex subunit 6|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6|NADH ubiquinone oxidoreductase B17 subunit|NADH ubiquinone oxidoreductase beta subunit 6|NADH-ubiquinone oxidoreductase B17 subunit|NDUB6_HUMAN|NDUFB6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95139
- O95139
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6 protein (2-67AA)
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6 protein (2-67AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together