Anti-GSTO2 Rabbit Polyclonal Antibody
Catalog # 77739-175
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GSTO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H4Y5
- Antigen Synonyms:GSTO2|Glutathione S-transferase omega-2|GSTO-2|EC 2.5.1.18|Glutathione S-transferase omega 2-2|GSTO 2-2|Glutathione-dependent dehydroascorbate reductase|EC 1.8.5.1|Monomethylarsonic acid reductase|MMA(V) reductase|EC 1.20.4.2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:EC 1.20.4.2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GSTO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat