Anti-MFNG Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MFNG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-074
- 77859-713
- Antigen Symbol:
- MFNG
- MFNG
- Antigen Synonyms:
- 3-N-acetylglucosaminyltransferase manic fringe|Beta-1|Beta-1,3-N-acetylglucosaminyltransferase manic fringe|MFNG_HUMAN|O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
- 3-N-acetylglucosaminyltransferase manic fringe|Beta-1|Beta-1,3-N-acetylglucosaminyltransferase manic fringe|MFNG_HUMAN|O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00587
- O00587
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Beta-1,3-N-acetylglucosaminyltransferase manic fringe protein (28-321AA)
- Recombinant Human Beta-1,3-N-acetylglucosaminyltransferase manic fringe protein (28-321AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together