Anti-MATK Rabbit Polyclonal Antibody
Catalog # 77742-819
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MATK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P42679
- Antigen Synonyms:CHK|Csk homologous kinase|Csk type protein tyrosine kinase|CTK|DKFZp434N1212|EC 2.7.10.2|Hematopoietic consensus tyrosine lacking kinase|Hematopoietic consensus tyrosine-lacking kinase|HHYLTK|Hydroxyaryl protein kinase|HYL|HYL tyrosine kinase|HYLTK|Leukocyte carboxyl terminal src kinase related|Lsk|MATK_HUMAN|Megakaryocyte associated tyrosine kinase|Megakaryocyte associated tyrosine protein kinase|Megakaryocyte-associated tyrosine-protein kinase|MGC1708|MGC2101|Protein kinase HYL|Tyrosine kinase Tyrosine protein kinase CTK|Tyrosine-protein kinase CTK|Tyrosylprotein kinase
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Tyrosylprotein kinase
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MATK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat