Anti-NLRP1 Rabbit Polyclonal Antibody
Catalog # 77744-509
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NLRP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:Q9C000
- Antigen Synonyms:CARD 7|CARD7|Caspase recruitment domain protein 7|Caspase recruitment domain-containing protein 7|CLR17.1|Death effector filament forming Ced 4 like apoptosis protein|Death effector filament-forming ced-4-like apoptosis protein|DEFCAP|DEFCAP L/S|DKFZp586O1822|KIAA0926|LRR and PYD domains-containing protein 1|NAC alpha/beta/gamma/delta|NAC|NACHT|NACHT leucine rich repeat and PYD containing 1|NACHT leucine rich repeat and PYD pyrin domain containing 1|NACHT leucine rich repeat and pyrin domain containing 1|NACHT LRR and PYD containing protein 1|NALP 1|NALP1|NALP1_HUMAN|NLR family pyrin domain containing 1|NLRP 1|NLRP1 protein|Nucleotide binding domain and caspase recruitment domain|Nucleotide binding oligomerization domain leucine rich repeat and pyrin domain containing 1|Nucleotide-binding domain and caspase recruitment domain|PP 1044|PP1044
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:PP1044
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NLRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human