Anti-FOLR2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: FOLR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-515
- 77737-375
- Antigen Symbol:
- FOLR2
- Antigen Synonyms:
- beta hFR|FBP|FBP/PL 1|FBP2|fetal/placental|folate receptor 2 (fetal)|Folate receptor 2|Folate receptor|Folate receptor beta|Folbp 2|Folbp2|Folr2|FOLR2_HUMAN|FR beta|FR P3|FR-beta|Placental folate binding protein|Placental folate-binding protein
- beta hFR|FBP|FBP/PL 1|FBP2|fetal/placental|folate receptor 2 (fetal)|Folate receptor 2|Folate receptor|Folate receptor beta|Folbp 2|Folbp2|Folr2|FOLR2_HUMAN|FR beta|FR P3|FR-beta|Placental folate binding protein|Placental folate-binding protein
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P14207
- P14207
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Immunogen:
- Recombinant Human Folate receptor beta protein (35-167AA)
- Recombinant Human Folate receptor beta protein (35-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together