Anti-INF2 Rabbit Polyclonal Antibody
77741-076
New Product
- Antigen Symbol:INF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q27J81
- Antigen Synonyms:C14orf151|C14orf173|CMTDIE|DKFZp762A0214|FLJ22056|FSGS5|HBEAG binding protein 2 binding protein C|HBEBP2 binding protein C|HBEBP2-binding protein C|INF 2|inf2|INF2_HUMAN|Inverted formin 2|Inverted formin FH2 and WH2 domain containing|Inverted formin-2|MGC13251|pp9484
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:pp9484
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: INF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat