Anti-Nox4 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB:1:500 to 1:5000
Type:
Antigen: Nox4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human
- Catalog No:
- 77744-723
- 77744-722
- Antigen Symbol:
- Nox4
- Nox4
- Antigen Synonyms:
- Nox4|Kox|NADPH oxidase 4|EC 1.6.3.-|Kidney oxidase-1|KOX-1|Kidney superoxide-producing NADPH oxidase
- Nox4|Kox|NADPH oxidase 4|EC 1.6.3.-|Kidney oxidase-1|KOX-1|Kidney superoxide-producing NADPH oxidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Rat, Human
- Rat, Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q924V1
- Q924V1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- NADPH oxidase 4 protein (210-424AA)
- NADPH oxidase 4 protein (210-424AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together