Anti-FDPS Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FDPS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-020
- 77855-283
- Antigen Symbol:
- FDPS
- FDPS
- Antigen Synonyms:
- (2E|(2E,6E) farnesyl diphosphate synthase|6E)-farnesyl diphosphate synthase|Dimethylallyltranstransferase|Farnesyl diphosphate synthase|Farnesyl diphosphate synthetase|Farnesyl pyrophosphate synthase|Farnesyl pyrophosphate synthetase|FPP synthase|FPP synthetase|FPPS|FPPS_HUMAN|FPS|Geranyltranstransferase
- (2E|(2E,6E) farnesyl diphosphate synthase|6E)-farnesyl diphosphate synthase|Dimethylallyltranstransferase|Farnesyl diphosphate synthase|Farnesyl diphosphate synthetase|Farnesyl pyrophosphate synthase|Farnesyl pyrophosphate synthetase|FPP synthase|FPP synthetase|FPPS|FPPS_HUMAN|FPS|Geranyltranstransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14324
- P14324
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Farnesyl pyrophosphate synthase protein (1-419AA)
- Recombinant Human Farnesyl pyrophosphate synthase protein (1-419AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together