Anti-FRS2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: FRS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-641
- 77855-616
- Antigen Symbol:
- FRS2
- FRS2
- Antigen Synonyms:
- FGFR signaling adaptor SNT|FGFR signalling adaptor|FGFR substrate 2|FGFR-signaling adaptor SNT|Fibroblast growth factor receptor substrate 2|FRS 2|FRS2_HUMAN|FRS2A|FRS2alpha|SNT 1|SNT|SNT-1|SNT1|Suc 1 Associated Neurotrophic Factor Target|Suc1 associated neurotrophic factor target 1|Suc1-associated neurotrophic factor target 1
- FGFR signaling adaptor SNT|FGFR signalling adaptor|FGFR substrate 2|FGFR-signaling adaptor SNT|Fibroblast growth factor receptor substrate 2|FRS 2|FRS2_HUMAN|FRS2A|FRS2alpha|SNT 1|SNT|SNT-1|SNT1|Suc 1 Associated Neurotrophic Factor Target|Suc1 associated neurotrophic factor target 1|Suc1-associated neurotrophic factor target 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WU20
- Q8WU20
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Fibroblast growth factor receptor substrate 2 protein (116-387AA)
- Recombinant human Fibroblast growth factor receptor substrate 2 protein (116-387AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together