Anti-IMPG2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: IMPG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-106
- 77741-071
- Antigen Symbol:
- IMPG2
- IMPG2
- Antigen Synonyms:
- IPM200|Interphotoreceptor matrix proteoglycan 2|Interphotoreceptor matrix proteoglycan of 200 kDa|IPM 200|Sialoprotein associated with cones and rods proteoglycan|Spacrcan
- IPM200|Interphotoreceptor matrix proteoglycan 2|Interphotoreceptor matrix proteoglycan of 200 kDa|IPM 200|Sialoprotein associated with cones and rods proteoglycan|Spacrcan
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZV3
- Q9BZV3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Interphotoreceptor matrix proteoglycan 2 protein (616-759AA)
- Recombinant human Interphotoreceptor matrix proteoglycan 2 protein (616-759AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together