Anti-MMP15 Rabbit Polyclonal Antibody
Catalog # 77739-541
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MMP15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P51511
- Antigen Synonyms:AI503551|Matrix metallopeptidase 15 membrane inserted|Matrix metalloproteinase 15|Matrix metalloproteinase-15|Membrane type 2 matrix metalloproteinase|Membrane type matrix metalloproteinase 2|Membrane-type matrix metalloproteinase 2|Membrane-type-2 matrix metalloproteinase|MMP 15|MMP-15|MMP15|MMP15_HUMAN|MT MMP 2|MT-MMP 2|MT2 MMP|MT2-MMP|MT2MMP|MTMMP 2|MTMMP2|SMCP 2|SMCP-2|SMCP2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:AI503551
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: MMP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse