Anti-MED6 Rabbit Polyclonal Antibody
Catalog # 77742-973
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MED6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O75586
- Antigen Synonyms:Activator recruited cofactor 33 kDa|Activator-recruited cofactor 33 kDa component|ARC33|hMED6_HUMAN|Mediator complex subunit 6|Mediator of RNA polymerase II transcription subunit 6|NY REN 28|NY REN 28 antigen|Renal carcinoma antigen NY-REN-28|RNA polymerase transcriptional regulation mediator subunit 6
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:RNA polymerase transcriptional regulation mediator subunit 6
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MED6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat