Anti-GCNT2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GCNT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-085
- 77855-870
- Antigen Symbol:
- GCNT2
- GCNT2
- Antigen Synonyms:
- II|N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase|IGNT|N-acetylglucosaminyltransferase|NACGT1|GCNT5|EC 2.4.1.150|I-branching enzyme
- GCNT5|II|NACGT1|N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase|N-acetylglucosaminyltransferase|EC 2.4.1.150|I-branching enzyme|IGNT
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N0V5
- Q8N0V5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase protein (201-402AA)
- Recombinant Human N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase protein (201-402AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together