Anti-ELF1 Rabbit Polyclonal Antibody
Catalog # 77735-798
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ELF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P32519
- Antigen Synonyms:E74 like factor 1|E74-like factor 1|ELF 1|ELF1|ELF1_HUMAN|Ets domain transcription factor|ETS related transcription factor Elf 1|ETS related transcription factor Elf1|ets-related transcription factor|ETS-related transcription factor Elf-1|mElf 1|p70|Sts1|TRANSCRIPTION FACTOR ELF 1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TRANSCRIPTION FACTOR ELF 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ELF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human