Anti-MT-CO1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MT-CO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-068
- 77743-560
- Antigen Symbol:
- MT-CO1
- MT-CO1
- Antigen Synonyms:
- COI|COX I|COX1|COX1_HUMAN|COXI|Cytochrome c oxidase polypeptide I|Cytochrome c oxidase subunit 1|Cytochrome C Oxidase subunit I|Mitochondrially encoded cytochrome c oxidase I|MT CO1|MTCO 1|MTCO1
- COI|COX I|COX1|COX1_HUMAN|COXI|Cytochrome c oxidase polypeptide I|Cytochrome c oxidase subunit 1|Cytochrome C Oxidase subunit I|Mitochondrially encoded cytochrome c oxidase I|MT CO1|MTCO 1|MTCO1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P00395
- P00395
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Cytochrome c oxidase subunit 1 protein (474-513AA)
- Recombinant Human Cytochrome c oxidase subunit 1 protein (474-513AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together