Anti-HSBP1 Rabbit Polyclonal Antibody
77740-404
New Product
- Antigen Symbol:HSBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O75506
- Antigen Synonyms:Heat shock factor binding protein 1|Heat shock factor-binding protein 1|HSBP1_HUMAN|HSF1BP|Nasopharyngeal carcinoma associated antigen 13|Nasopharyngeal carcinoma-associated antigen 13|NPC A 13|NPC-A-13|NPCA 13
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:NPCA 13
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: HSBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat