Anti-NISCH Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NISCH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-458
- 77744-459
- Antigen Symbol:
- NISCH
- NISCH
- Antigen Synonyms:
- Imidazoline-1 receptor candidate protein|Imidazoline receptor 1|I1R candidate protein|I-1 receptor candidate protein|IR1|Nischarin|I1R|I-1|KIAA0975|Imidazoline receptor antisera-selected protein|hIRAS|IRAS|Imidazoline-1 receptor
- Imidazoline-1 receptor candidate protein|Imidazoline receptor 1|I1R candidate protein|I-1 receptor candidate protein|IR1|Nischarin|I1R|I-1|KIAA0975|Imidazoline receptor antisera-selected protein|hIRAS|IRAS|Imidazoline-1 receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2I1
- Q9Y2I1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nischarin protein (500-700AA)
- Nischarin protein (500-700AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together