Order Entry
United States
ContactUsLinkComponent
Anti-MSX2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MSX2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MSX2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77860-064
  • 77743-556
  • Antigen Symbol:
  • MSX2
  • MSX2
  • Antigen Synonyms:
  • CRS 2|CRS2|FPP|Homeo box msh like 2|Homeobox protein Hox-8|Homeobox protein MSX 2|Homeobox protein MSX-2|Homeobox protein Hox 8|Hox8|MSH|Msh homeo box 2|Msh homeo box homolog|Msh homeo box homolog 2|Msh homeobox 2|Msh homeobox homolog 2|Msx 2|MSX2_HUMAN|Parietal foramina 1|PFM 1|PFM|PFM1
  • CRS 2|CRS2|FPP|Homeo box msh like 2|Homeobox protein Hox-8|Homeobox protein MSX 2|Homeobox protein MSX-2|Homeobox protein Hox 8|Hox8|MSH|Msh homeo box 2|Msh homeo box homolog|Msh homeo box homolog 2|Msh homeobox 2|Msh homeobox homolog 2|Msx 2|MSX2_HUMAN|Parietal foramina 1|PFM 1|PFM|PFM1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P35548
  • P35548
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Homeobox protein MSX-2 protein (50-120AA)
  • Recombinant Human Homeobox protein MSX-2 protein (50-120AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together