Anti-NXF1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: NXF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-262
- 77745-263
- Antigen Symbol:
- NXF1
- NXF1
- Antigen Synonyms:
- DKFZp667O0311|DmMEX67|MEX67, yeast, homolog of|Mex67p|mRNA export factor TAP|Mvb1|Nuclear RNA export factor 1|Nuclear RNA export factor 1 homolog (S. cerevisiae)|nxf 1|NXF1_HUMAN|Protein small bristles|Sbr|TAP|Tip associating protein|Tip-associated protein|Tip-associating protein
- DKFZp667O0311|DmMEX67|MEX67, yeast, homolog of|Mex67p|mRNA export factor TAP|Mvb1|Nuclear RNA export factor 1|Nuclear RNA export factor 1 homolog (S. cerevisiae)|nxf 1|NXF1_HUMAN|Protein small bristles|Sbr|TAP|Tip associating protein|Tip-associated protein|Tip-associating protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBU9
- Q9UBU9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Nuclear RNA export factor 1 protein (1-130AA)
- Nuclear RNA export factor 1 protein (1-130AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together