Anti-RAD54B Rabbit Polyclonal Antibody
Catalog # 77737-027
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RAD54B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y620
- Antigen Synonyms:DNA repair and recombination protein EC 3.6.4.-|RAD54 homolog B
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:RAD54 homolog B
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: RAD54B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human