Anti-NCBP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NCBP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-987
- 77743-988
- Antigen Symbol:
- NCBP2
- NCBP2
- Antigen Synonyms:
- CBP20|20 kDa nuclear cap-binding protein|NCBP-interacting protein 1|PIG55|NIP1|Nuclear cap-binding protein subunit 2|Cell proliferation-inducing gene 55 protein|NCBP 20 kDa subunit
- CBP20|20 kDa nuclear cap-binding protein|NCBP-interacting protein 1|PIG55|NIP1|Nuclear cap-binding protein subunit 2|Cell proliferation-inducing gene 55 protein|NCBP 20 kDa subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52298
- P52298
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nuclear cap-binding protein subunit 2 protein (1-156AA)
- Nuclear cap-binding protein subunit 2 protein (1-156AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together