Anti-ELK3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: ELK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77854-685
- 77735-818
- Antigen Symbol:
- ELK3
- ELK3
- Antigen Synonyms:
- ETS domain protein (SRF accessory protein 2)|ELK3_HUMAN|ERP|ETS domain protein|ETS domain-containing protein Elk-3|ETS related protein ERP|ETS related protein NET|ETS-related protein ERP|ETS-related protein NET|NET|SAP 2|SAP-2|SAP2|Serum response factor accessory protein 2|SRF accessory protein 2
- ETS related protein ERP|ERP|SAP 2|ETS domain protein|ETS-related protein ERP|ETS domain protein (SRF accessory protein 2)|ETS-related protein NET|ELK3_HUMAN|Serum response factor accessory protein 2|SAP2|SAP-2|ETS related protein NET|NET|ETS domain-containing protein Elk-3|SRF accessory protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41970
- P41970
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human ETS domain-containing protein Elk-3 protein (163-300AA)
- Recombinant Human ETS domain-containing protein Elk-3 protein (163-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together