Anti-HIP1R Rabbit Polyclonal Antibody
Catalog # 77739-694
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HIP1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O75146
- Antigen Synonyms:HIP3|Huntingtin-interacting protein 1-related protein|HIP1R_HUMAN|HIP12|ILWEQ|Huntingtin-interacting protein 12|FLJ27022|MGC47513|Hip1 related|KIAA0655|Huntingtin interacting protein 12|FLJ14000|HIP1-related protein|HIP-12|Huntingtin Interacting Protein 1 Related
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MGC47513
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HIP1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat