Anti-MCHR1 Rabbit Polyclonal Antibody
Catalog # 77742-885
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MCHR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q99705
- Antigen Synonyms:GPR24|SLC1Melanin-concentrating hormone receptor 1|MCH receptor 1|MCH-R1|MCHR-1|G-protein coupled receptor 24|MCH-1R|MCH1R|MCHR|SLC-1|Somatostatin receptor-like protein
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Somatostatin receptor-like protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MCHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat