Anti-GLUD2 Rabbit Polyclonal Antibody
77738-459
New Product
- Antigen Symbol:GLUD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P49448
- Antigen Synonyms:DHE4_HUMAN| GDH 2| GDH| GDH2| GLUDP1| Glutamate dehydrogenase 2| Glutamate dehydrogenase 2 mitochondrial| Glutamate dehydrogenase pseudogene 1| mitochondrial
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:mitochondrial
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: GLUD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat