Anti-ENC1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: ENC1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-727
- 77735-894
- Antigen Symbol:
- ENC1
- ENC1
- Antigen Synonyms:
- CCL 28|CCL28|Ectoderm neural cortex protein 1|Ectoderm-neural cortex protein 1|Ectodermal neural cortex (with BTB like domain)|ENC 1|ENC-1|ENC1_HUMAN|FLJ39259|Kelch like 35|Kelch like 37|Kelch-like protein 37|KLHL 35|KLHL35|KLHL37|NRPB|Nuclear matrix protein NRP/B|Nuclear restricted protein BTB domain like (brain)|OTTHUMP00000128235|OTTHUMP00000222054|p53 induced gene 10 protein|p53-induced gene 10 protein|PIG 10|PIG10|TP53I10|Tumor protein p53 inducible protein 10
- CCL 28| CCL28| Ectoderm neural cortex protein 1| Ectoderm-neural cortex protein 1| Ectodermal neural cortex (with BTB like domain)| ENC 1| ENC-1| ENC1_HUMAN| FLJ39259| Kelch like 35| Kelch like 37| Kelch-like protein 37| KLHL 35| KLHL35| KLHL37| NRPB| Nuclear matrix protein NRP/B| Nuclear restricted protein BTB domain like (brain)| OTTHUMP00000128235| OTTHUMP00000222054| p53 induced gene 10 protein| p53-induced gene 10 protein| PIG 10| PIG10| TP53I10| Tumor protein p53 inducible protein 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14682
- O14682
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ectoderm-neural cortex protein 1 protein (1-230AA)
- Recombinant Human Ectoderm-neural cortex protein 1 protein (1-230AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together