Anti-GGPS1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GGPS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-959
- 77738-210
- Antigen Symbol:
- GGPS1
- GGPS1
- Antigen Synonyms:
- (2E|6E)-farnesyl diphosphate synthase|Dimethylallyltranstransferase|Farnesyl diphosphate synthase|Farnesyltranstransferase|Geranylgeranyl diphosphate synthase 1|Geranylgeranyl diphosphate synthase|Geranylgeranyl pyrophosphate synthase|Geranylgeranyl pyrophosphate synthetase|Geranyltranstransferase|GGPP synthase|GGPP synthetase|GGPPS|GGPPS_HUMAN|GGPPS1|GGPPSase|OTTHUMP00000036073|OTTHUMP00000216161|OTTHUMP00000216163|OTTHUMP00000216164
- (2E|6E)-farnesyl diphosphate synthase|Dimethylallyltranstransferase|Farnesyl diphosphate synthase|Farnesyltranstransferase|Geranylgeranyl diphosphate synthase 1|Geranylgeranyl diphosphate synthase|Geranylgeranyl pyrophosphate synthase|Geranylgeranyl pyrophosphate synthetase|Geranyltranstransferase|GGPP synthase|GGPP synthetase|GGPPS|GGPPS_HUMAN|GGPPS1|GGPPSase|OTTHUMP00000036073|OTTHUMP00000216161|OTTHUMP00000216163|OTTHUMP00000216164
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95749
- O95749
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Geranylgeranyl pyrophosphate synthase protein (1-300AA)
- Recombinant Human Geranylgeranyl pyrophosphate synthase protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together