Anti-IRF4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: IRF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-189
- 77741-183
- Antigen Symbol:
- IRF4
- IRF4
- Antigen Synonyms:
- Interferon regulatory factor 4|IRF 4|IRF-4|IRF4_HUMAN|LSIRF|Lymphocyte specific interferon regulatory factor|Lymphocyte specific IRF|Lymphocyte-specific interferon regulatory factor|Multiple myeloma oncogene 1|MUM 1|MUM1|NF EM5|NF-EM5|NFEM5|PU.1 interaction partner|Sfpi1/PU.1 interaction partner|Transcriptional activator PIP
- Interferon regulatory factor 4|IRF 4|IRF-4|IRF4_HUMAN|LSIRF|Lymphocyte specific interferon regulatory factor|Lymphocyte specific IRF|Lymphocyte-specific interferon regulatory factor|Multiple myeloma oncogene 1|MUM 1|MUM1|NF EM5|NF-EM5|NFEM5|PU.1 interaction partner|Sfpi1/PU.1 interaction partner|Transcriptional activator PIP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15306
- Q15306
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon regulatory factor 4 protein (180-451AA)
- Recombinant Human Interferon regulatory factor 4 protein (180-451AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together