Anti-MAP3K5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP3K5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-678
- 77859-403
- Antigen Symbol:
- MAP3K5
- MAP3K5
- Antigen Synonyms:
- Apoptosis signal regulating kinase 1|Apoptosis signal-regulating kinase 1|ASK 1|ASK-1|ASK1|M3K5|M3K5_HUMAN|MAP/ERK kinase kinase 5|MAPK/ERK kinase kinase 5|MAPKKK5|MEK kinase 5|MEKK 5|MEKK5|Mitogen activated protein kinase kinase kinase 5|Mitogen-activated protein kinase kinase kinase 5
- Apoptosis signal regulating kinase 1|Apoptosis signal-regulating kinase 1|ASK 1|ASK-1|ASK1|M3K5|M3K5_HUMAN|MAP/ERK kinase kinase 5|MAPK/ERK kinase kinase 5|MAPKKK5|MEK kinase 5|MEKK 5|MEKK5|Mitogen activated protein kinase kinase kinase 5|Mitogen-activated protein kinase kinase kinase 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99683
- Q99683
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase kinase kinase 5 protein (363-431AA)
- Recombinant Human Mitogen-activated protein kinase kinase kinase 5 protein (363-431AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together