Anti-IPO13 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IPO13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-165
- 77741-156
- Antigen Symbol:
- IPO13
- IPO13
- Antigen Synonyms:
- Imp 13|Imp13|Importin-13|Importin13|IPO 13|IPO13_HUMAN|Kap 13|Kap13|Karyopherin 13|Karyopherin-13|Karyopherin13|KIAA0274|Late gestation lung 2|Late gestation lung 2 protein|Lgl2|Ran binding protein 13|Ran-binding protein 13|RanBP 13|RanBP13
- IPO13_HUMAN|Importin13|Lgl2|Importin-13|IPO 13|Karyopherin 13|Late gestation lung 2 protein|KIAA0274|Kap 13|RanBP 13|Ran-binding protein 13|Kap13|Imp 13|Karyopherin13|Late gestation lung 2|Imp13|Ran binding protein 13|Karyopherin-13|RanBP13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94829
- O94829
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Importin-13 protein (501-700AA)
- Recombinant Human Importin-13 protein (501-700AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together