Anti-MAP4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-702
- 77859-423
- Antigen Symbol:
- MAP4
- MAP4
- Antigen Synonyms:
- DKFZp779A1753|MAP-4|MAP4_HUMAN|MGC8617|Microtubule associated protein 4|Microtubule-associated protein 4|OTTHUMP00000210723|OTTHUMP00000210725|OTTHUMP00000210727|OTTHUMP00000210730
- DKFZp779A1753|MAP-4|MAP4_HUMAN|MGC8617|Microtubule associated protein 4|Microtubule-associated protein 4|OTTHUMP00000210723|OTTHUMP00000210725|OTTHUMP00000210727|OTTHUMP00000210730
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27816
- P27816
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Microtubule-associated protein 4 protein (575-818AA)
- Recombinant Human Microtubule-associated protein 4 protein (575-818AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together