Anti-NCOA1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NCOA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-040
- 77744-039
- Antigen Symbol:
- NCOA1
- NCOA1
- Antigen Synonyms:
- bHLHe74|Class E basic helix-loop-helix protein 74|F SRC 1|Hin 2 protein|Hin2 protein|MGC129719|MGC129720|mNRC 1|NCoA 1|NCoA-1|NCOA1_HUMAN|Nuclear receptor coactivator 1|Nuclear receptor coactivator protein 1|NY REN 52 antigen|Protein Hin 2|Protein Hin-2|Protein Hin2|Renal carcinoma antigen NY REN 52|Renal carcinoma antigen NY-REN-52|RIP 160|RIP160|SRC 1|SRC-1|Steroid receptor coactivator 1
- bHLHe74|Class E basic helix-loop-helix protein 74|F SRC 1|Hin 2 protein|Hin2 protein|MGC129719|MGC129720|mNRC 1|NCoA 1|NCoA-1|NCOA1_HUMAN|Nuclear receptor coactivator 1|Nuclear receptor coactivator protein 1|NY REN 52 antigen|Protein Hin 2|Protein Hin-2|Protein Hin2|Renal carcinoma antigen NY REN 52|Renal carcinoma antigen NY-REN-52|RIP 160|RIP160|SRC 1|SRC-1|Steroid receptor coactivator 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15788
- Q15788
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear receptor coactivator 1 protein (284-393AA)
- Nuclear receptor coactivator 1 protein (284-393AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together