Anti-HNRNPM Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: HNRNPM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-543
- 77740-254
- Antigen Symbol:
- HNRNPM
- HNRNPM
- Antigen Synonyms:
- CEA receptor|CEAR|Heterogeneous nuclear ribonucleoprotein M|Heterogenous nuclear ribonucleoprotein M4|hnRNA binding protein M4|hnRNP M|HNRNPM4|HNRPM|HNRPM_HUMAN|HNRPM4|HTGR1|M3|M4|M4 protein|N-acetylglucosamine receptor 1|NAGR1
- CEA receptor|CEAR|Heterogeneous nuclear ribonucleoprotein M|Heterogenous nuclear ribonucleoprotein M4|hnRNA binding protein M4|hnRNP M|HNRNPM4|HNRPM|HNRPM_HUMAN|HNRPM4|HTGR1|M3|M4|M4 protein|N-acetylglucosamine receptor 1|NAGR1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52272
- P52272
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Heterogeneous nuclear ribonucleoprotein M protein (491-730AA)
- Recombinant Human Heterogeneous nuclear ribonucleoprotein M protein (491-730AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together