Anti-MAP2K7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: MAP2K7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77742-652
- 77859-380
- Antigen Symbol:
- MAP2K7
- MAP2K7
- Antigen Synonyms:
- JNKK 2|SAPK kinase 4|stress-activated protein kinase kinase 4|MAP kinase kinase 7|MAPKK7|JNK kinase 2|MAPKK 7|SKK4|SAPKK4|Sek 2|MP2K7_HUMAN|MKK7|SAPKK-4|MEK 7|PRKMK 7|PRKMK7|Mitogen Activated Protein Kinase kinase 7|Sek-2|Dual specificity mitogen activated protein kinase kinase 7|MAPK/ERK kinase 7|MKK 7|Jnkk2|JNK-activating kinase 2|c-Jun N-terminal kinase kinase 2|Jnkk-2|MAPKK-7|Dual specificity mitogen-activated protein kinase kinase 7|PRKMK-7|JNK activating kinase 2|MKK-7|Sek2
- c-Jun N-terminal kinase kinase 2|Dual specificity mitogen activated protein kinase kinase 7|Dual specificity mitogen-activated protein kinase kinase 7|JNK activating kinase 2|JNK kinase 2|JNK-activating kinase 2|JNKK 2|Jnkk-2|Jnkk2|MAP kinase kinase 7|MAPK/ERK kinase 7|MAPKK 7|MAPKK-7|MAPKK7|MEK 7|Mitogen Activated Protein Kinase kinase 7|MKK 7|MKK-7|MKK7|MP2K7_HUMAN|PRKMK 7|PRKMK-7|PRKMK7|SAPK kinase 4|SAPKK-4|SAPKK4|Sek 2|Sek-2|Sek2|SKK4|stress-activated protein kinase kinase 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14733
- O14733
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 7 protein (17-160AA)
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 7 protein (17-160AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together